α-Bromo-2-chlorotoulene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-(Bromomethyl)-2-chlorobenzene, 2-Chlorobenzyl bromide
M-Clarity™ quality level = MQ100
252255-25G 252255-5G
SIAL252255-25GEA 131 EUR
SIAL252255-25G SIAL252255-5G
α-Bromo-2-chlorotoulene, Sigma-Aldrich®
α-Bromo-2-chlorotoulene
Formula: C₇H₆BrCl
Boiling Pt: 235 °C (1013 hPa)
Density: 1.5897 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000566
CAS Number: 611-17-6
EINECS: 210-257-2
UN: 3265
ADR: 8,II