α-Bromo-3-chlorotoulene, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: α-Bromo-m-chlorotoulene, 3-Chlorobenzyl bromide, 1-(Bromomethyl)-3-chlorobenzene, alpha-Bromo-3-chlorotoluene

M-Clarity™ quality level = MQ100

136727-10G 136727-50G
SIAL136727-10GEA 41 EUR
SIAL136727-10G SIAL136727-50G
α-Bromo-3-chlorotoulene, Sigma-Aldrich®
α-Bromo-3-chlorotoulene
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 17.5 °C
Density: 1.56 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000597
CAS Number: 766-80-3
EINECS: 212-171-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR