α-Bromo-4-chlorotoulene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: alpha-Bromo-4-chlorotoluene, 1-(Bromomethyl)-4-chlorobenzene, 4-Chlorobenzyl bromide
M-Clarity™ quality level = MQ200
558958-100G 558958-25G
SIAL558958-100GEA 913 EUR
SIAL558958-100G SIAL558958-25G
α-Bromo-4-chlorotoulene, Sigma-Aldrich®
α-Bromo-4-chlorotoulene
Formula: C₇H₆BrCl
Boiling Pt: 118…123 °C (19 mmHg)
Melting Pt: 48…50 °C
MDL Number: MFCD00040714
CAS Number: 622-95-7
EINECS: 210-760-7
UN: 3261
ADR: 8,II