(2-Bromophenyl)acetonitrile, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Bromobenzyl cyanide
M-Clarity™ quality level = MQ200
259969-25G 259969-5G
SIAL259969-25GEA 59.8 EUR
SIAL259969-25G SIAL259969-5G
(2-Bromophenyl)acetonitrile, Sigma-Aldrich®
(2-Bromophenyl)acetonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 1 °C
Density: 1.51 g/cm³ (20 °C)
MDL Number: MFCD00001896
CAS Number: 19472-74-3
EINECS: 243-091-4
UN: 1694
ADR: 6.1,I