ω-Bromoacetophenone, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
α-Bromoacetophenone, Phenacyl bromide, 2-Bromoacetophenone
M-Clarity™ quality level = MQ100
115835-100G
115835-500G
115835-5G
SIAL115835-100GEA
79
EUR
SIAL115835-100G
SIAL115835-500G
SIAL115835-5G
ω-Bromoacetophenone, Sigma-Aldrich®
ω-Bromoacetophenone
Formula:
(C₆H₅)C(O)CH₂Br MW: 199.05 g/mol Boiling Pt: 253 °C (1013 hPa) Melting Pt: 50 °C Density: 1.647 g/cm³ (20 °C) Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000195 CAS Number: 70-11-1 EINECS: 200-724-9 UN: 2645 ADR: 6.1,II Merck Index: 13,01386 |