2'-Bromoacetophenone, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
o-Bromoacetophenone
M-Clarity™ quality level = MQ100
183695-25G
183695-5G
SIAL183695-25GEA
126
EUR
SIAL183695-25G
SIAL183695-5G
2'-Bromoacetophenone, Sigma-Aldrich®
2'-Bromoacetophenone
Formula:
C₈H₇BrO MW: 199.05 g/mol Boiling Pt: 249 °C (1013 hPa) Melting Pt: 0 °C Density: 1.476 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000067 CAS Number: 2142-69-0 EINECS: 218-398-1 |