4'-Bromoacetophenone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
M-Clarity™ quality level = MQ100
B56404-100G B56404-25G
SIALB56404-100GEA 70.4 EUR
SIALB56404-100G SIALB56404-25G
4'-Bromoacetophenone, Sigma-Aldrich®
4'-Bromoacetophenone
Formula: C₈H₇BrO
MW: 199.05 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 49…52 °C
Density: 1.736 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000105
CAS Number: 99-90-1
EINECS: 202-799-3
UN: 2811
ADR: 6.1,II
Merck Index: 13,01387