ω-Bromo-4-methoxyacetophenone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 4-Methoxyphenacyl bromide, Bromomethyl p-methoxyphenyl ketone, α-Bromo-4-methoxyacetophenone, 2-Bromo-1-(4-methoxyphenyl)ethan-1-one, 2-Bromo-1-(4-methoxyphenyl)ethanone, 2-Bromo-4'-methoxyacetophenone
M-Clarity™ quality level = MQ100
115665-100G 115665-25G 115665-5G
SIAL115665-100GEA 668 EUR
SIAL115665-100G SIAL115665-25G SIAL115665-5G
ω-Bromo-4-methoxyacetophenone, Sigma-Aldrich®
ω-Bromo-4-methoxyacetophenone
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 285
Melting Pt: 67…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000201
CAS Number: 2632-13-5
EINECS: 220-118-8
UN: 3261
ADR: 8 II