α-Bromo-2-fluorotoulene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Fluorobenzyl bromide, α-Bromo-o-fluorotoulene

M-Clarity™ quality level = MQ100

209511-25G 209511-5G
SIAL209511-25GEA 81.8 EUR
SIAL209511-25G SIAL209511-5G
α-Bromo-2-fluorotoulene, Sigma-Aldrich®
α-Bromo-2-fluorotoulene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 84 °C (1013 hPa)
Density: 1.567 g/cm³ (20 °C)
MDL Number: MFCD00000324
CAS Number: 446-48-0
EINECS: 207-169-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR