3,5-Difluorobenzyl bromide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: α-Bromo-3,5-difluorotoluene
M-Clarity™ quality level = MQ100
290319-25G 290319-5G
SIAL290319-25GEA 120 EUR
SIAL290319-25G SIAL290319-5G
3,5-Difluorobenzyl bromide, Sigma-Aldrich®
3,5-Difluorobenzyl bromide
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 65 °C (4.5 mmHg)
Density: 1.6 g/cm³ (20 °C)
MDL Number: MFCD00010304
CAS Number: 141776-91-2
UN: 3265
ADR: 8,II