α-Bromo-4-fluorotoulene, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
α-Bromo-p-fluorotoulene, 4-Fluorobenzyl bromide, p-Fluorobenzyl bromide
M-Clarity™ quality level = MQ100
209538-25G
209538-5G
SIAL209538-25GEA
105
EUR
SIAL209538-25G
SIAL209538-5G
α-Bromo-4-fluorotoulene, Sigma-Aldrich®
α-Bromo-4-fluorotoulene
Formula:
C₇H₆BrF MW: 189.03 g/mol Boiling Pt: 85 °C (1013 hPa) Density: 1.517 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000359 CAS Number: 459-46-1 EINECS: 207-291-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...