2-Bromo-α,α,α-trifluorotoluene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Bromo-2-(trifluoromethyl)benzene, 2-Bromobenzotrifluoride
M-Clarity™ quality level = MQ200
B58806-100G B58806-25G
SIALB58806-100GEA 88.5 EUR
SIALB58806-100G SIALB58806-25G
2-Bromo-α,α,α-trifluorotoluene, Sigma-Aldrich®
2-Bromo-α,α,α-trifluorotoluene
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 167 °C (1013 hPa)
Density: 1.651 g/cm³ (30 °C)
MDL Number: MFCD00000373
CAS Number: 392-83-6
EINECS: 206-879-9
UN: 1993
ADR: 3,III