2-Bromo-5-fluorotoluene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Bromo-4-fluoro-2-methylbenzene
M-Clarity™ quality level = MQ100
365009-25G 365009-5G
SIAL365009-25GEA 31 EUR
SIAL365009-25G SIAL365009-5G
2-Bromo-5-fluorotoluene, Sigma-Aldrich®
2-Bromo-5-fluorotoluene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 177 °C (1013 hPa)
Density: 1.49 g/cm³ (20 °C)
MDL Number: MFCD00017921
CAS Number: 452-63-1
EINECS: 207-203-5
UN: 1993
ADR: 3,III