3-Bromo-5-(trifluoromethyl)aniline, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
3-Amino-5-bromo-α,α,α-trifluorotoluene, 5-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-5-bromobenzotrifluoride
M-Clarity™ quality level = MQ100
539813-25G
539813-5G
SIAL539813-25GEA
89.2
EUR
SIAL539813-25G
SIAL539813-5G
3-Bromo-5-(trifluoromethyl)aniline, Sigma-Aldrich®
3-Bromo-5-(trifluoromethyl)aniline
Formula:
C₇H₅BrF₃N MW: 240.02 g/mol |
MDL Number:
MFCD00236205 CAS Number: 54962-75-3 EINECS: 259-412-6 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...