α-Bromo-4-iodotoulene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 4-Iodobenzyl bromide, 1-(Bromomethyl)-4-iodobenzene, α-Bromo-p-iodotoulene
M-Clarity™ quality level = MQ100
515604-1G 515604-5G
SIAL515604-1GEA 28.3 EUR
SIAL515604-1G SIAL515604-5G
α-Bromo-4-iodotoulene, Sigma-Aldrich®
α-Bromo-4-iodotoulene
Formula: C₇H₆BrI
MW: 296.93 g/mol
Boiling Pt: 115…120 °C (0.6 mmHg)
Melting Pt: 80
Storage Temperature: Refrigerator
MDL Number: MFCD00209656
CAS Number: 16004-15-2
UN: 3261
ADR: 8 II