2-Bromo-p-toluidine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Bromo-4-methylaniline, 4-Amino-3-bromotoluene
M-Clarity™ quality level = MQ100
124087-10G 124087-25G
SIAL124087-10GEA 77.4 EUR
SIAL124087-10G SIAL124087-25G
2-Bromo-p-toluidine, Sigma-Aldrich®
2-Bromo-p-toluidine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 26 °C
Density: 1.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007635
CAS Number: 583-68-6
EINECS: 209-515-7
UN: 2810
ADR: 6.1,III