α-Bromo-4-nitrotoluene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 4-Nitrobenzyl bromide, 1-(Bromomethyl)-4-nitrobenzene, α-Bromo-p-nitrotoluene
M-Clarity™ quality level = MQ200
N13054-100G N13054-25G
SIALN13054-100GEA 59.8 EUR
SIALN13054-100G SIALN13054-25G
α-Bromo-4-nitrotoluene, Sigma-Aldrich®
α-Bromo-4-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 304
Melting Pt: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00007373
CAS Number: 100-11-8
EINECS: 202-820-6
UN: 3261
ADR: 8 II