2-Bromo-p-cresol
Supplier: Thermo Scientific
Warning
Synonyms: 2-Bromo-4-methylphenol, 1-Bromo-2-hydroxy-5-methylbenzene
SB01962.1 SB01962.10 SB01962.50
MAYBSB01962.1EA 0 EUR
MAYBSB01962.1 MAYBSB01962.10 MAYBSB01962.50
2-Bromo-p-cresol
2-Bromo-p-cresol
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 213
Melting Pt: 26
Density: 1.5468 g/cm³ (25 °C)
MDL Number: MFCD00002151
CAS Number: 6627-55-0
EINECS: 229-595-7