2-Bromo-p-cresol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Bromo-4-methylphenol, 1-Bromo-2-hydroxy-5-methylbenzene
M-Clarity™ quality level = MQ100
B72809-25G B72809-5G
SIALB72809-25GEA 198 EUR
SIALB72809-25G SIALB72809-5G
2-Bromo-p-cresol, Sigma-Aldrich®
2-Bromo-p-cresol
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 26 °C
Density: 1.5468 g/cm³ (25 °C)
MDL Number: MFCD00002151
CAS Number: 6627-55-0
EINECS: 229-595-7