4-Isopropylbenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Cuminaldehyde
W234109-1KG-K
W234109-5KG-K
SIALW234109-1KG-KEA
141
EUR
SIALW234109-1KG-K
SIALW234109-5KG-K
4-Isopropylbenzaldehyde, Sigma-Aldrich®
4-Isopropylbenzaldehyde
Formula:
C₁₀H₁₂O MW: 148.2 g/mol |
MDL Number:
MFCD00006953 CAS Number: 122-03-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...