4-Methoxy-3-methylbenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Methyl-4-anisaldehyde
M-Clarity™ quality level = MQ100
152129-10G 152129-50G
SIAL152129-10GEA 238 EUR
SIAL152129-10G SIAL152129-50G
4-Methoxy-3-methylbenzaldehyde, Sigma-Aldrich®
4-Methoxy-3-methylbenzaldehyde
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 80
Density: 1.082 g/cm³ (16 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003375
CAS Number: 32723-67-4
EINECS: 251-177-8