5-Methylsalicylaldehyde, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
2-Hydroxy-5-methylbenzaldehyde
454664-1G
454664-5G
SIAL454664-1GEA
54.4
EUR
SIAL454664-1G
SIAL454664-5G
5-Methylsalicylaldehyde, Sigma-Aldrich®
5-Methylsalicylaldehyde
Formula:
C₈H₈O₂ MW: 136.15 g/mol Boiling Pt: 217 °C (1013 hPa) Melting Pt: 54 °C Density: 1,0913 g/cm³ (59 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00274297 CAS Number: 613-84-3 EINECS: 210-357-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...