(3-Chloropropyl)benzene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Chloro-3-phenylpropane, 3-Phenylpropyl chloride, 3-Phenylpropyl Chloride
M-Clarity™ quality level = MQ100
C68105-100ML C68105-5ML
SIALC68105-100MLEA 96.8 EUR
SIALC68105-100ML SIALC68105-5ML
(3-Chloropropyl)benzene, Sigma-Aldrich®
(3-Chloropropyl)benzene
Formula: ClH₂CCH₂CH₂(C₆H₅)
MW: 154.64 g/mol
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: –27 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001001
CAS Number: 104-52-9
EINECS: 203-210-2