2,4-Dichlorotoluene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,3-Dichloro-4-methylbenzene
M-Clarity™ quality level = MQ100
145009-1KG 145009-250G
SIAL145009-1KGEA 148 EUR
SIAL145009-1KG SIAL145009-250G
2,4-Dichlorotoluene, Sigma-Aldrich®
2,4-Dichlorotoluene
Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: –13 °C
Density: 1.2498 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00000583
CAS Number: 95-73-8
EINECS: 202-445-8
UN: 3082
ADR: 9,III