(3,4-Dichlorophenyl)acetonitrile, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3,4-Dichlorophenylacetonitrile, 3,4-Dichlorobenzyl cyanide

M-Clarity™ quality level = MQ200

232475-100G 232475-25G
SIAL232475-100GEA 125 EUR
SIAL232475-100G SIAL232475-25G
(3,4-Dichlorophenyl)acetonitrile, Sigma-Aldrich®
(3,4-Dichlorophenyl)acetonitrile
Formula: C₈H₅Cl₂N
MW: 186.04 g/mol
Boiling Pt: 309 °C (1013 hPa)
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00001909
CAS Number: 3218-49-3
EINECS: 221-743-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR