3',4'-Dichloroacetophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-(3,4-dichlorophenyl)ethanone
160520-100G
160520-25G
SIAL160520-100GEA
67.5
EUR
SIAL160520-100G
SIAL160520-25G
3',4'-Dichloroacetophenone, Sigma-Aldrich®
3',4'-Dichloroacetophenone
Formula:
C₈H₆Cl₂O MW: 189.04 g/mol Boiling Pt: 265 °C (1013 hPa) Melting Pt: 44…46 °C |
MDL Number:
MFCD00000553 CAS Number: 2642-63-9 EINECS: 220-146-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...