2',6'-Dichloroacetophenone ≥98%
Supplier: Thermo Scientific
Warning
Synonyms: 1-(2,6-Dichlorophenyl)ethanone
L06348.03 L06348.06
L06348.03EA 60.5 EUR
L06348.03 L06348.06
2',6'-Dichloroacetophenone ≥98%
2',6'-Dichloroacetophenone
Purity: 98% CAS No.: 2040-05-3 Molecular Formula: C8H6Cl2O Formula Weight: 189.04 Melting Point: 41-44° Boiling Point: 176-180°/83mm MDL No.: MFCD00016339
Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 176…180 °C (83 mmHg)
Melting Pt: 41…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00016339
CAS Number: 2040-05-3
EINECS: 218-035-7