2',5'-Dichloroacetophenone ≥98%
Supplier: Thermo Fisher Scientific
Warning
A13770.09 A13770.14 A13770.22
A13770.09EA 39.1 EUR
A13770.09 A13770.14 A13770.22
2',5'-Dichloroacetophenone ≥98%
2',5'-Dichloroacetophenone
Purity: 98% CAS No.: 2476-37-1 Molecular Formula: C8H6Cl2O Formula Weight: 189.04 Flash Point: 110°(230°F) Refractive Index: 1.5610 Melting Point: 11-13° Boiling Point: 118°/12mm Beilstein Reference: 1865043 MDL No.: MFCD00000607
Formula: C₈H₆Cl₂O
Boiling Pt: 118 °C (12 mmHg)
Melting Pt: 11…13 °C
Density: 1.312
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000607
CAS Number: 2476-37-1
EINECS: 219-605-8