ω,3,4-Trichloroacetophenone
Supplier: Thermo Fisher Scientific
Danger
Synonyms: α,3,4-Trichloroacetophenone, 2-Chloro-1-(3,4-dichlorophenyl)ethanone, 3,4-Dichlorophenacyl chloride, 2,3',4'-Trichloroacetophenone
B25397.06 B25397.14
B25397.06EA 73.3 EUR
B25397.06 B25397.14
ω,3,4-Trichloroacetophenone
ω,3,4-Trichloroacetophenone
CAS No.: 42981-08-8 Molecular Formula: C8H5Cl3O Formula Weight: 223.49 Melting Point: 43-45° Boiling Point: 171-173°/15mm UN No.: 1759 MDL No.: MFCD03970382
Formula: C₈H₅Cl₃O
MW: 223.49 g/mol
Boiling Pt: 171…173 °C (15 mmHg)
Melting Pt: 43…45 °C
Storage Temperature: Ambient
MDL Number: MFCD03970382
CAS Number: 42981-08-8
UN: 1759
ADR: 8 III