2'-Chloroacetophenone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-(2-Chlorophenyl)ethanone, o-Chloroacetophenone
M-Clarity™ quality level = MQ100
183709-100G 183709-25G
SIAL183709-100GEA 59.8 EUR
SIAL183709-100G SIAL183709-25G
2'-Chloroacetophenone, Sigma-Aldrich®
2'-Chloroacetophenone
Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 227 °C (1013 hPa)
Density: 1.189 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000560
CAS Number: 2142-68-9
EINECS: 218-397-6
UN: 3416
ADR: 6.1,II