4'-Chloropropiophenone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-(4-Chlorophenyl)propan-1-one

M-Clarity™ quality level = MQ100

C69209-100G C69209-25G
SIALC69209-100GEA 115 EUR
SIALC69209-100G SIALC69209-25G
4'-Chloropropiophenone, Sigma-Aldrich®
4'-Chloropropiophenone
Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 277 °C (1013 hPa)
Melting Pt: 33…34 °C
MDL Number: MFCD00000626
CAS Number: 6285-05-8
EINECS: 228-511-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR