3-Chloro-p-toluic acid

Supplier: Thermo Scientific
Warning

TL00693DA TL00693EA
MAYBTL00693DAEA 0 EUR
MAYBTL00693DA MAYBTL00693EA
3-Chloro-p-toluic acid
3-Chloro-p-toluic acid
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
MDL Number: MFCD00045841
CAS Number: 5162-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR