4-Chloro-o-toluic acid

Supplier: Thermo Scientific
Warning

TL00910DA TL00910EA
MAYBTL00910DAEA 0 EUR
MAYBTL00910DA MAYBTL00910EA
4-Chloro-o-toluic acid
4-Chloro-o-toluic acid
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
MDL Number: MFCD00045852
CAS Number: 7499-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR