α-Chloro-2,6-difluorotoulene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-(Chloromethyl)-1,3-difluorobenzene, 2,6-Difluorobenzyl chloride, 1-(Chloromethyl)-2,6-difluorobenzene
M-Clarity™ quality level = MQ100
559954-25G 559954-5G
SIAL559954-25GEA 263 EUR
SIAL559954-25G SIAL559954-5G
α-Chloro-2,6-difluorotoulene, Sigma-Aldrich®
α-Chloro-2,6-difluorotoulene
Formula: C₇H₅ClF₂
MW: 162.57 g/mol
Boiling Pt: 172…173 °C (640 mmHg)
Melting Pt: 32 °C
MDL Number: MFCD00191346
CAS Number: 697-73-4
UN: 1759
ADR: 8,II