4-Fluorobenzyl chloride ≥99%, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 4-Fluorobenzyl chloride, p-Fluorobenzyl chloride, 1-Chloromethyl-4-fluorobenzene, α-Chloro-4-fluorotoluene, 1-(Chloromethyl)-4-fluorobenzene
M-Clarity™ quality level = MQ100
F8004-100G-A F8004-25G-A
SIALF8004-100G-AEA 58.7 EUR
SIALF8004-100G-A SIALF8004-25G-A
4-Fluorobenzyl chloride ≥99%, Sigma-Aldrich®
4-Fluorobenzyl chloride
Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 181.2 °C (1013 hPa)
Melting Pt: –18 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00000913
CAS Number: 352-11-4
EINECS: 206-516-4
UN: 3265
ADR: 8,II