2-Chloro-4-(trifluoromethyl)aniline, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
578568-25G 578568-5G
SIAL578568-25GEA 49.7 EUR
SIAL578568-25G SIAL578568-5G
2-Chloro-4-(trifluoromethyl)aniline, Sigma-Aldrich®
2-Chloro-4-(trifluoromethyl)aniline
Formula: C₇H₅ClF₃N
Boiling Pt: 89 °C (0.5 mmHg)
MDL Number: MFCD00042563
CAS Number: 39885-50-2
EINECS: 254-674-8
UN: 2810
ADR: 6.1,III