4-Chloro-3-(trifluoromethyl)phenyl isocyanate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL374881-5GEA 95.9 EUR
SIAL374881-5G
4-Chloro-3-(trifluoromethyl)phenyl isocyanate, Sigma-Aldrich®
4-Chloro-3-(trifluoromethyl)phenyl isocyanate
Formula: C₈H₃ClF₃NO
MW: 221,57 g/mol
Boiling Pt: 86…90 °C (14 mmHg)
Melting Pt: 27,2…27.5 °C
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR