2,3-Dichlorobenzylamine 97%
Supplier: Thermo Fisher Scientific
|
Danger
|
L11884.03
L11884.06
L11884.14
L11884.03EA
33.9
EUR
L11884.03
L11884.06
L11884.14
2,3-Dichlorobenzylamine 97%
2,3-Dichlorobenzylamine
Purity: 97%
CAS No.: 39226-95-4
Molecular Formula: C7H7Cl2N
Formula Weight: 176.05
Flash Point: 105°(221°F)
Refractive Index: 1.5820
Boiling Point: 124-126°/15mm
Beilstein Reference: 2639296
UN No.: 2735
MDL No.: MFCD00047927
Formula:
C₇H₇Cl₂N Boiling Pt: 124…126 °C (15 mmHg) Density: 1.321 Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00047927 CAS Number: 39226-95-4 UN: 2735 ADR: 8,III |