2,4-Dichlorobenzylamine 98%
Supplier: Thermo Fisher Scientific
Danger
113230100 113230500
ACRO113230100EA 38.4 EUR
ACRO113230100 ACRO113230500
2,4-Dichlorobenzylamine 98%
2,4-Dichlorobenzylamine
Formula: C₇H₇Cl₂N
Boiling Pt: 83 °C (1013 hPa)
Density: 1.32 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00008109
CAS Number: 95-00-1
EINECS: 202-382-6
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5760 to 1.5800 (20°C, 589 nm)
GC ≥97.5 %
Suitability (for use as a laser dye) Passes test
Molar absorption coefficient ≥77000 l/(mol.cm) (at Lambda max)