2,4-Dichlorobenzylamine 98%
Supplier: Thermo Scientific
Danger
Synonyms: (2,4-Dichlorophenyl)methanamine
A13207.06 A13207.14
A13207.06EA 36.2 EUR
A13207.06 A13207.14
2,4-Dichlorobenzylamine 98%
2,4-Dichlorobenzylamine
Purity: 98% CAS No.: 95-00-1 Molecular Formula: C7H7Cl2N Formula Weight: 176.05 Flash Point: 110°(230°F) Refractive Index: 1.5780 Odour: Amine-like Boiling Point: 258-260° Beilstein Reference: 775667 UN No.: 2735 MDL No.: MFCD00008109
Formula: C₇H₇Cl₂N
MW: 176.04 g/mol
Boiling Pt: 258…260 °C
Density: 1.320
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008109
CAS Number: 95-00-1
EINECS: 202-382-6
UN: 2735
ADR: 8,III