2-Chlorobenzylamine
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(2-Chlorophenyl)methanamine
SB01016.1
SB01016.10
SB01016.50
MAYBSB01016.1EA
0
EUR
MAYBSB01016.1
MAYBSB01016.10
MAYBSB01016.50
2-Chlorobenzylamine
2-Chlorobenzylamine
Formula:
C₇H₈ClN MW: 141.6 g/mol Boiling Pt: 103 °C (1013 hPa) Density: 1.17 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008108 CAS Number: 89-97-4 EINECS: 201-955-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...