2-Chlorobenzylamine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: (2-Chlorophenyl)methanamine
M-Clarity™ quality level = MQ100
C27204-100G C27204-25G
SIALC27204-100GEA 114 EUR
SIALC27204-100G SIALC27204-25G
2-Chlorobenzylamine, Sigma-Aldrich®
2-Chlorobenzylamine
Formula: C₇H₈ClN
Boiling Pt: 103 °C (1013 hPa)
Density: 1.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008108
CAS Number: 89-97-4
EINECS: 201-955-8
UN: 2735
ADR: 8,III