3,5-Dichlorobenzylamine
Supplier: Thermo Scientific
Danger
TL00168DA TL00168EA
MAYBTL00168DAEA 0 EUR
MAYBTL00168DA MAYBTL00168EA
3,5-Dichlorobenzylamine
3,5-Dichlorobenzylamine
Formula: C₇H₇Cl₂N
MW: 176.04 g/mol
Boiling Pt: 254 °C (6 mmHg)
Density: 1.32 g/cm³ (20 °C)
MDL Number: MFCD00052681
CAS Number: 39989-43-0
UN: 2735
ADR: 8,III