3-Chloro-p-toluidine ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
3-Chloro-p-methylaniline, 2-Chloro-4-aminotoluene, 4-Amino-2-chlorotoluene, 3-Chloro-4-methylaniline
A15439.0B
A15439.30
A15439.0BEA
247
EUR
A15439.0B
A15439.30
3-Chloro-p-toluidine ≥98%
3-Chloro-p-toluidine
Purity: 98%
CAS No.: 95-74-9
Molecular Formula: C7H8ClN
Formula Weight: 141.60
Flash Point: 130°(266°F)
Refractive Index: 1.5840
Melting Point: 22-24°
Boiling Point: 236-238°
Beilstein Reference: 636511
UN No.: 2239
MDL No.: MFCD00007773
Formula:
C₇H₈ClN MW: 141.6 g/mol Boiling Pt: 236…238 °C Melting Pt: 22…24 °C Density: 1.180 Flash Pt: 130 °C (266 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007773 CAS Number: 95-74-9 EINECS: 202-446-3 UN: 2239 ADR: 6.1 III |