α-Chloro-4-nitrotoluene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Nitrobenzyl chloride, 4-Nitrobenzylchloride, α-Chloro-p-nitrotoluene
M-Clarity™ quality level = MQ100
140112-100G 140112-25G
SIAL140112-100GEA 195 EUR
SIAL140112-100G SIAL140112-25G
α-Chloro-4-nitrotoluene, Sigma-Aldrich®
α-Chloro-4-nitrotoluene
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 293
Melting Pt: 68…70 °C
Density: 1.5647 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007374
CAS Number: 100-14-1
EINECS: 202-822-7
UN: 3261
ADR: 8 II