6-Chloro-o-cresol
Supplier: Thermo Scientific
Synonyms: 1-Chloro-2-hydroxy-3-methylbenzene, 2-Chloro-6-methylphenol
SB00441.1 SB00441.10 SB00441.50
MAYBSB00441.1EA 0 EUR
MAYBSB00441.1 MAYBSB00441.10 MAYBSB00441.50
6-Chloro-o-cresol
6-Chloro-o-cresol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.1966 g/cm³ (20 °C)
MDL Number: MFCD00020000
CAS Number: 87-64-9
EINECS: 201-760-8
UN: 2669
ADR: 6.1,III