6-Chloro-m-cresol ≥99%
Supplier: Thermo Scientific
Danger
Synonyms: 1-Chloro-2-hydroxy-4-methylbenzene, 2-Chloro-5-methylphenol
L00816.14 L00816.22
L00816.14EA 53 EUR
L00816.14 L00816.22
6-Chloro-m-cresol ≥99%
6-Chloro-m-cresol
Purity: 99% CAS No.: 615-74-7 Molecular Formula: C7H7ClO Formula Weight: 142.59 Flash Point: 81°(177°F) Melting Point: 44-48° Boiling Point: 195-197° Beilstein Reference: 2041487 UN No.: 3437 MDL No.: MFCD00002175
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 195…197 °C
Melting Pt: 44…48 °C
Density: 1.215
Flash Pt: 81 °C (177 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002175
CAS Number: 615-74-7
EINECS: 210-444-9
UN: 3437
ADR: 6.1,II