2-Chloro-p-cresol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1-Chloro-2-hydroxy-5-methylbenzene, 2-Chloro-4-methylphenol

M-Clarity™ quality level = MQ100

273198-25G 273198-5G
SIAL273198-25GEA 229 EUR
SIAL273198-25G SIAL273198-5G
2-Chloro-p-cresol, Sigma-Aldrich®
2-Chloro-p-cresol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 198…203 °C (1013 hPa)
Melting Pt: 76 °C
Density: 1.211 g/cm³ (25 °C)
MDL Number: MFCD00070639
CAS Number: 6640-27-3
EINECS: 229-656-8
UN: 2669
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR