3-Chlorobenzyl alcohol

Supplier: Thermo Scientific

Synonyms: (3-Chlorophenyl)methanol

SB01130.1 SB01130EA SB01130EE
MAYBSB01130.1EA 0 EUR
MAYBSB01130.1 MAYBSB01130EA MAYBSB01130EE
3-Chlorobenzyl alcohol
3-Chlorobenzyl alcohol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 237 °C (1013 hPa)
Density: 1.21 g/cm³ (20 °C)
MDL Number: MFCD00004632
CAS Number: 873-63-2
EINECS: 212-847-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR