5-Chloro-o-cresol ≥98%
Supplier: Thermo Scientific
Danger
Synonyms: 1-Chloro-3-hydroxy-4-methylbenzene, 5-Chloro-2-methylphenol
B25527.06 B25527.14
B25527.06EA 77.2 EUR
B25527.06 B25527.14
5-Chloro-o-cresol ≥98%
5-Chloro-o-cresol
Purity: 98% CAS No.: 5306-98-9 Molecular Formula: C7H7ClO Formula Weight: 142.59 Melting Point: 70-75° Boiling Point: 109-112°/15mm UN No.: 3437 MDL No.: MFCD00060672
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 109…112 °C (15 mmHg)
Melting Pt: 70…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00060672
CAS Number: 5306-98-9
EINECS: 226-160-3
UN: 3437
ADR: 6.1,II